Assay Details
| Gene Symbol: Gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853640.2 | XP_004853697.1 | 2 - 3 | 305 | 77 | |
| XM_004853641.3 | XP_004853698.1 | 3 - 4 | 354 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853641.3 transcripts.
Set Membership: |
|

Gnai3
Genomic Map