Assay Details
| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855421.3 | XP_004855478.1 | 2 - 3 | 2144 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo3| Gene Symbol: Foxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855421.3 | XP_004855478.1 | 2 - 3 | 2144 | 75 | |
Set Membership: |
|