Assay Details
| Gene Symbol: Tbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856087.2 | XP_004856144.1 | 4 - 5 | 1127 | 67 | |
| XM_013076319.2 | XP_012931773.1 | 5 - 6 | 925 | 67 | ||
More Information
Set Membership: |
|

Tbx6
Genomic Map