Assay Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865139.3 | XP_004865196.1 | 4 - 5 | 977 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
