Assay Details
| Gene Symbol: Plcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871686.2 | XP_004871743.1 | 4 - 5 | 2779 | 65 | |
| XM_004871690.3 | XP_004871747.1 | 4 - 5 | 2831 | 65 | ||
| XM_021249788.1 | XP_021105447.1 | 4 - 5 | 2937 | 65 | ||
| XM_021249789.1 | XP_021105448.1 | 4 - 5 | 2779 | 65 | ||
| XM_021249790.1 | XP_021105449.1 | 4 - 5 | 2779 | 65 | ||
| XM_021249791.1 | XP_021105450.1 | 4 - 5 | 2816 | 65 | ||
More Information
Set Membership: |
|

Plcl1
Genomic Map