Assay Details
| Gene Symbol: Gpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873890.2 | XP_004873947.1 | 3 - 4 | 656 | 59 | |
| XM_021251472.1 | XP_021107131.1 | 3 - 4 | 241 | 59 | ||
| XM_021251473.1 | XP_021107132.1 | 3 - 4 | 213 | 59 | ||
More Information
Set Membership: |
|

Gpt2
Genomic Map