Assay Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077799.2 | XP_012933253.1 | 3 - 4 | 401 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tac4| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077799.2 | XP_012933253.1 | 3 - 4 | 401 | 84 | |