Assay Details
| Gene Symbol: LOC101712848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247517.1 | XP_021103176.1 | 15 - 16 | 2086 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101712848| Gene Symbol: LOC101712848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247517.1 | XP_021103176.1 | 15 - 16 | 2086 | 73 | |
Set Membership: |
|