Assay Details
| Gene Symbol: Maf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256683.1 | XP_021112342.1 | 3 - 4 | 2455 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maf| Gene Symbol: Maf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256683.1 | XP_021112342.1 | 3 - 4 | 2455 | 80 | |
Set Membership: |
|