Assay Details
| Gene Symbol: LOC101700566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265103.1 | XP_021120762.1 | 3 - 4 | 1211 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101700566| Gene Symbol: LOC101700566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265103.1 | XP_021120762.1 | 3 - 4 | 1211 | 56 | |
Set Membership: |
|