Assay Details
| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845897.1 | XP_004845954.1 | 5 - 6 | 764 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnpla5| Gene Symbol: Pnpla5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845897.1 | XP_004845954.1 | 5 - 6 | 764 | 58 | |
Set Membership: |
|