Assay Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847355.3 | XP_004847412.1 | 5 - 6 | 1431 | 65 | |
| XM_004847356.3 | XP_004847413.1 | 4 - 5 | 1398 | 65 | ||
| XM_021260359.1 | XP_021116018.1 | 5 - 6 | 1437 | 65 | ||
More Information
Set Membership: |
|

Cpz
Genomic Map