Assay Details
| Gene Symbol: Stpg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848739.3 | XP_004848796.1 | 1 - 2 | 1074 | 55 | |
| XM_004848741.3 | XP_004848798.1 | 1 - 2 | 1073 | 55 | ||
| XM_021261609.1 | XP_021117268.1 | 1 - 2 | 1074 | 55 | ||
| XM_021261610.1 | XP_021117269.1 | 1 - 2 | 1073 | 55 | ||
More Information
Set Membership: |
|

Stpg3
Genomic Map