Assay Details
| Gene Symbol: Stpg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848739.3 | XP_004848796.1 | 3 - 4 | 1347 | 79 | |
| XM_004848741.3 | XP_004848798.1 | 3 - 4 | 1346 | 79 | ||
| XM_021261609.1 | XP_021117268.1 | 3 - 4 | 1347 | 79 | ||
| XM_021261610.1 | XP_021117269.1 | 3 - 4 | 1346 | 79 | ||
More Information
Set Membership: |
|

Stpg3
Genomic Map