Assay Details
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854729.2 | XP_004854786.1 | 4 - 5 | 1312 | 106 | |
| XM_021266467.1 | XP_021122126.1 | 4 - 5 | 3135 | 106 | ||
| XM_021266468.1 | XP_021122127.1 | 5 - 6 | 2253 | 106 | ||
| XM_021266469.1 | XP_021122128.1 | 5 - 6 | 1362 | 106 | ||
| XM_021266471.1 | XP_021122130.1 | 4 - 5 | 3109 | 106 | ||
| XM_021266472.1 | XP_021122131.1 | 4 - 5 | 1713 | 106 | ||
| XM_021266473.1 | XP_021122132.1 | 3 - 4 | 2888 | 106 | ||
More Information
Set Membership: |
|

Smad9
Genomic Map