Assay Details
| Gene Symbol: Jpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860646.3 | XP_004860703.1 | 3 - 4 | 443 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jpt1| Gene Symbol: Jpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860646.3 | XP_004860703.1 | 3 - 4 | 443 | 86 | |