Assay Details
| Gene Symbol: Arsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870278.3 | XP_004870335.1 | 11 - 12 | 2189 | 68 | |
| XM_021248616.1 | XP_021104275.1 | 12 - 13 | 2502 | 68 | ||
| XM_021248617.1 | XP_021104276.1 | 13 - 14 | 2629 | 68 | ||
| XM_021248618.1 | XP_021104277.1 | 12 - 13 | 3306 | 68 | ||
| XM_021248619.1 | XP_021104278.1 | 12 - 13 | 2410 | 68 | ||
| XM_021248620.1 | XP_021104279.1 | 9 - 10 | 2155 | 68 | ||
| XM_021248621.1 | XP_021104280.1 | 11 - 12 | 1435 | 68 | ||
| XM_021248622.1 | XP_021104281.1 | 7 - 8 | 1084 | 68 | ||
| XM_021248623.1 | XP_021104282.1 | 7 - 8 | 972 | 68 | ||
| XM_021248624.1 | XP_021104283.1 | 6 - 7 | 931 | 68 | ||
More Information
Set Membership: |
|

Arsg
Genomic Map