Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870285.3 | XP_004870342.1 | 3 - 4 | 497 | 72 | |
| XM_004870286.3 | XP_004870343.1 | 3 - 4 | 469 | 72 | ||
| XM_021248655.1 | XP_021104314.1 | 4 - 5 | 604 | 72 | ||
More Information
Set Membership: |
|

Prkar1a
Genomic Map