Assay Details
| Gene Symbol: Ofd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870996.3 | XP_004871053.1 | 10 - 11 | 1231 | 69 | |
| XM_013067884.2 | XP_012923338.1 | 10 - 11 | 1269 | 69 | ||
| XM_013067887.2 | XP_012923341.1 | 10 - 11 | 1231 | 69 | ||
| XM_013067888.2 | XP_012923342.1 | 7 - 8 | 780 | 69 | ||
| XM_013067889.2 | XP_012923343.1 | 8 - 9 | 844 | 69 | ||
| XM_021249122.1 | XP_021104781.1 | 10 - 11 | 1176 | 69 | ||
| XM_021249123.1 | XP_021104782.1 | 10 - 11 | 1228 | 69 | ||
| XM_021249124.1 | XP_021104783.1 | 9 - 10 | 1030 | 69 | ||
| XM_021249125.1 | XP_021104784.1 | 9 - 10 | 1068 | 69 | ||
More Information
Set Membership: |
|

Ofd1
Genomic Map