Assay Details
| Gene Symbol: LOC101723875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238442.1 | XP_021094101.1 | 5 - 6 | 682 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101723875| Gene Symbol: LOC101723875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021238442.1 | XP_021094101.1 | 5 - 6 | 682 | 86 | |