Assay Details
| Gene Symbol: LOC101720880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245710.1 | XP_021101369.1 | 12 - 13 | 1732 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101720880| Gene Symbol: LOC101720880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245710.1 | XP_021101369.1 | 12 - 13 | 1732 | 81 | |
Set Membership: |
|