Assay Details
| Gene Symbol: LOC101720880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245710.1 | XP_021101369.1 | 1 - 2 | 251 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245710.1 transcripts.
Set Membership: |
|

LOC101720880
Genomic Map