Assay Details
| Gene Symbol: Znf496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252125.1 | XP_021107784.1 | 7 - 8 | 1375 | 79 | |
| XM_021252127.1 | XP_021107786.1 | 7 - 8 | 1925 | 79 | ||
| XM_021252128.1 | XP_021107787.1 | 7 - 8 | 1758 | 79 | ||
| XM_021252129.1 | XP_021107788.1 | 7 - 8 | 1310 | 79 | ||
More Information
Set Membership: |
|

Znf496
Genomic Map