Assay Details
| Gene Symbol: CUNH3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261515.1 | XP_021117174.1 | 1 - 2 | 62 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021261515.1 transcripts.
Set Membership: |
|

CUNH3orf84
Genomic Map