Assay Details
| Gene Symbol: CUNH3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261515.1 | XP_021117174.1 | 3 - 4 | 238 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH3orf84| Gene Symbol: CUNH3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261515.1 | XP_021117174.1 | 3 - 4 | 238 | 57 | |
Set Membership: |
|