Assay Details
| Gene Symbol: CUNH3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265153.1 | XP_021120812.1 | 1 - 2 | 1223 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH3orf30| Gene Symbol: CUNH3orf30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021265153.1 | XP_021120812.1 | 1 - 2 | 1223 | 117 | |
Set Membership: |
|