Assay Details
| Gene Symbol: Rab36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843531.3 | XP_004843588.1 | 3 - 4 | 291 | 67 | |
| XM_004843532.3 | XP_004843589.1 | 3 - 4 | 291 | 67 | ||
| XM_004843535.3 | XP_004843592.1 | 3 - 4 | 292 | 67 | ||
| XM_013071688.2 | XP_012927142.1 | 3 - 4 | 325 | 67 | ||
More Information
Set Membership: |
|

Rab36
Genomic Map