Assay Details
| Gene Symbol: Pcdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844283.2 | XP_004844340.1 | 4 - 5 | 4057 | 82 | |
| XM_004844284.2 | XP_004844341.1 | 3 - 4 | 3954 | 82 | ||
| XM_004844285.2 | XP_004844342.1 | 4 - 5 | 3931 | 82 | ||
| XM_004844286.2 | XP_004844343.1 | 3 - 4 | 3828 | 82 | ||
| XM_021257934.1 | XP_021113593.1 | 4 - 5 | 3464 | 82 | ||
More Information
Set Membership: |
|

Pcdh9
Genomic Map