Assay Details
| Gene Symbol: Papd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845158.3 | XP_004845215.1 | 7 - 8 | 1202 | 67 | |
| XM_004845162.3 | XP_004845219.1 | 7 - 8 | 1197 | 67 | ||
| XM_004845163.3 | XP_004845220.1 | 7 - 8 | 983 | 67 | ||
| XM_021258597.1 | XP_021114256.1 | 7 - 8 | 1198 | 67 | ||
More Information
Set Membership: |
|

Papd7
Genomic Map