Assay Details
| Gene Symbol: Pnpla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846687.2 | XP_004846744.1 | 2 - 3 | 816 | 99 | |
| XM_021259921.1 | XP_021115580.1 | 2 - 3 | 816 | 99 | ||
| XM_021259922.1 | XP_021115581.1 | 2 - 3 | 816 | 99 | ||
| XM_021259923.1 | XP_021115582.1 | 2 - 3 | 816 | 99 | ||
| XM_021259924.1 | XP_021115583.1 | 2 - 3 | 816 | 99 | ||
| XM_021259926.1 | XP_021115585.1 | 2 - 3 | 816 | 99 | ||
| XM_021259927.1 | XP_021115586.1 | 2 - 3 | 816 | 99 | ||
| XM_021259928.1 | XP_021115587.1 | 2 - 3 | 816 | 99 | ||
More Information
Set Membership: |
|

Pnpla1
Genomic Map