Assay Details
| Gene Symbol: CUNH6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847166.2 | XP_004847223.1 | 1 - 2 | 1709 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH6orf15| Gene Symbol: CUNH6orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847166.2 | XP_004847223.1 | 1 - 2 | 1709 | 69 | |
Set Membership: |
|