Assay Details
| Gene Symbol: Selenop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848565.3 | XP_004848622.2 | 3 - 4 | 643 | 120 | |
| XM_013073510.2 | XP_012928964.1 | 3 - 4 | 643 | 120 | ||
More Information
Set Membership: |
|

Selenop
Genomic Map