Assay Details
| Gene Symbol: Anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854043.3 | XP_004854100.1 | 9 - 10 | 1863 | 66 | |
| XM_013075485.2 | XP_012930939.1 | 9 - 10 | 1867 | 66 | ||
| XM_013075486.2 | XP_012930940.1 | 8 - 9 | 1177 | 66 | ||
| XM_013075487.2 | XP_012930941.1 | 8 - 9 | 1783 | 66 | ||
| XM_013075488.2 | XP_012930942.1 | 8 - 9 | 1241 | 66 | ||
| XM_021265924.1 | XP_021121583.1 | 7 - 8 | 1091 | 66 | ||
More Information
Set Membership: |
|

Anxa9
Genomic Map