Assay Details
| Gene Symbol: LOC101717715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859426.2 | XP_004859483.1 | 6 - 7 | 1182 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101717715| Gene Symbol: LOC101717715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859426.2 | XP_004859483.1 | 6 - 7 | 1182 | 126 | |
Set Membership: |
|