Assay Details
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862433.2 | XP_004862490.1 | 3 - 4 | 194 | 82 | |
XM_004862435.3 | XP_004862492.2 | 2 - 3 | 265 | 82 | ||
XM_013078719.2 | XP_012934173.2 | 3 - 4 | 209 | 82 | ||
XM_021242619.1 | XP_021098278.1 | 3 - 4 | 250 | 82 | ||
XM_021242620.1 | XP_021098279.1 | 3 - 4 | 194 | 82 | ||
XM_021242622.1 | XP_021098281.1 | 3 - 4 | 273 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021242622.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
