Assay Details
| Gene Symbol: Bicra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867217.3 | XP_004867274.1 | 14 - 15 | 3843 | 55 | |
| XM_021246017.1 | XP_021101676.1 | 14 - 15 | 4036 | 55 | ||
| XM_021246018.1 | XP_021101677.1 | 14 - 15 | 3273 | 55 | ||
| XM_021246020.1 | XP_021101679.1 | 13 - 14 | 3099 | 55 | ||
More Information
Set Membership: |
|

Bicra
Genomic Map