Assay Details
| Gene Symbol: Bicra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867217.3 | XP_004867274.1 | 7 - 8 | 2512 | 112 | |
| XM_021246017.1 | XP_021101676.1 | 7 - 8 | 2705 | 112 | ||
More Information
Set Membership: |
|

Bicra
Genomic Map