Assay Details
| Gene Symbol: Npas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867578.3 | XP_004867635.2 | 7 - 8 | 1031 | 59 | |
| XM_004867579.3 | XP_004867636.2 | 6 - 7 | 947 | 59 | ||
| XM_013066627.2 | XP_012922081.1 | 7 - 8 | 904 | 59 | ||
| XM_013066630.2 | XP_012922084.2 | 6 - 7 | 947 | 59 | ||
| XM_013066631.2 | XP_012922085.1 | 6 - 7 | 834 | 59 | ||
| XM_021246622.1 | XP_021102281.1 | 7 - 8 | 1037 | 59 | ||
| XM_021246624.1 | XP_021102283.1 | 7 - 8 | 1037 | 59 | ||
| XM_021246625.1 | XP_021102284.1 | 8 - 9 | 1088 | 59 | ||
More Information
Set Membership: |
|

Npas3
Genomic Map