Assay Details
| Gene Symbol: Borcs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873425.3 | XP_004873482.1 | 5 - 6 | 545 | 89 | |
| XM_021251205.1 | XP_021106864.1 | 5 - 6 | 637 | 89 | ||
More Information
Set Membership: |
|

Borcs8
Genomic Map