Assay Details
| Gene Symbol: Rab6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065020.2 | XP_012920474.1 | 7 - 8 | 784 | 82 | |
| XM_013065021.2 | XP_012920475.1 | 7 - 8 | 784 | 82 | ||
More Information
Set Membership: |
|

Rab6a
Genomic Map