Assay Details
| Gene Symbol: Creb3l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065783.2 | XP_012921237.1 | 7 - 8 | 1398 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Creb3l3| Gene Symbol: Creb3l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065783.2 | XP_012921237.1 | 7 - 8 | 1398 | 69 | |
Set Membership: |
|