Assay Details
| Gene Symbol: Ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260635.1 | XP_021116294.1 | 10 - 11 | 1202 | 66 | |
| XM_021260636.1 | XP_021116295.1 | 11 - 12 | 1444 | 66 | ||
| XM_021260637.1 | XP_021116296.1 | 12 - 13 | 1725 | 66 | ||
More Information
Set Membership: |
|

Ranbp9
Genomic Map