Assay Details
| Gene Symbol: LOC101696934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840231.2 | XP_004840288.2 | 5 - 6 | 1061 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696934| Gene Symbol: LOC101696934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840231.2 | XP_004840288.2 | 5 - 6 | 1061 | 65 | |
Set Membership: |
|