Assay Details
| Gene Symbol: LOC101725754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843076.3 | XP_004843133.1 | 2 - 3 | 364 | 95 | |
| XM_004843077.3 | XP_004843134.1 | 2 - 3 | 365 | 95 | ||
| XM_013071580.2 | XP_012927034.1 | 2 - 3 | 366 | 95 | ||
More Information
Set Membership: |
|

LOC101725754
Genomic Map