Assay Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862064.3 | XP_004862121.1 | 5 - 6 | 1127 | 66 | |
| XM_004862066.3 | XP_004862123.1 | 4 - 5 | 1000 | 66 | ||
More Information
Set Membership: |
|

Rassf5
Genomic Map