Assay Details
| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864556.3 | XP_004864613.1 | 8 - 9 | 1823 | 57 | |
| XM_004864557.3 | XP_004864614.1 | 8 - 9 | 819 | 57 | ||
More Information
Set Membership: |
|

Cnppd1
Genomic Map