Assay Details
| Gene Symbol: Ptchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866191.2 | XP_004866248.1 | 2 - 3 | 966 | 63 | |
| XM_021245318.1 | XP_021100977.1 | 1 - 2 | 586 | 63 | ||
| XM_021245319.1 | XP_021100978.1 | 2 - 3 | 896 | 63 | ||
More Information
Set Membership: |
|

Ptchd1
Genomic Map