Assay Details
| Gene Symbol: Pdcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868513.3 | XP_004868570.1 | 2 - 3 | 5373 | 62 | |
| XM_004868514.3 | XP_004868571.1 | 2 - 3 | 5376 | 62 | ||
| XM_004868515.3 | XP_004868572.1 | 3 - 4 | 689 | 62 | ||
| XM_004868516.3 | XP_004868573.1 | 2 - 3 | 5378 | 62 | ||
More Information
Set Membership: |
|

Pdcd1
Genomic Map