Assay Details
| Gene Symbol: Mapk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868778.1 | XP_004868835.1 | 5 - 6 | 2409 | 70 | |
| XM_004868780.1 | XP_004868837.1 | 3 - 4 | 2006 | 70 | ||
| XM_021247523.1 | XP_021103182.1 | 5 - 6 | 2981 | 70 | ||
| XM_021247524.1 | XP_021103183.1 | 5 - 6 | 2412 | 70 | ||
| XM_021247525.1 | XP_021103184.1 | 4 - 5 | 2295 | 70 | ||
| XM_021247526.1 | XP_021103185.1 | 4 - 5 | 1915 | 70 | ||
| XM_021247527.1 | XP_021103186.1 | 3 - 4 | 1885 | 70 | ||
More Information
Set Membership: |
|

Mapk7
Genomic Map