Assay Details
| Gene Symbol: Nrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872081.2 | XP_004872138.1 | 5 - 6 | 452 | 130 | |
| XM_021250093.1 | XP_021105752.1 | 5 - 6 | 449 | 130 | ||
| XM_021250095.1 | XP_021105754.1 | 4 - 5 | 360 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872081.2, XM_021250093.1, XM_021250095.1 transcripts.
Set Membership: |
|

Nrg4
Genomic Map